Gene Transcripts

Gene Symbol
Krt28
Entrez Gene ID
360623
Gene Name
keratin 28, type I
Gene Aliases
Ka41, Krt25d
Chromosome Location
Chr. 10: 87276096 - 87286618 on Build Rnor_6.0
UniGene ID
Rn.105748
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008760.1 NP_001008760.1 7 1045
XM_006247356.2 XP_006247418.1 8 1513

View Full Product Details