Gene Transcripts

Gene Symbol
Krt84
Entrez Gene ID
315320
Gene Name
keratin 84, type II
Gene Aliases
Kb24
Chromosome Location
Chr. 7: 143180229 - 143187663 on Build Rnor_6.0
UniGene ID
Rn.143278
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008812.1 NP_001008812.1 4 869
XM_006242346.2 XP_006242408.1 4 938

View Full Product Details