Gene Transcripts

Gene Symbol
Pfn4
Entrez Gene ID
494222
Gene Name
profilin family, member 4
Gene Aliases
Chromosome Location
Chr. 6: 30027915 - 30038000 on Build Rnor_6.0
UniGene ID
Rn.126513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009503.1 NP_001009503.1 4 1305
GenBank mRNA AY682392.1 - 4 1305
BC107465.1 - 4 1437 7453892

View Full Product Details