Gene Transcripts

Gene Symbol
Clec4d
Entrez Gene ID
362432
Gene Name
C-type lectin domain family 4, member D
Gene Aliases
Clecsf8, Mcl
Chromosome Location
Chr. 4: 156253084 - 156262644 on Build Rnor_6.0
UniGene ID
Rn.144379
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003707.1 NP_001003707.1 6 647
XM_006237322.2 XP_006237384.1 6 829
GenBank mRNA AY339882.1 - 6 647
AY494065.1 - 6 647

View Full Product Details