Gene Transcripts

Gene Symbol
G6b
Entrez Gene ID
406865
Gene Name
immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor
Gene Aliases
C6orf25, Ng31
Chromosome Location
Chr. 20: 5054421 - 5057026 on Build Rnor_6.0
UniGene ID
Rn.142706
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003976.1 NP_001003976.1 5 685
XM_006256069.2 XP_006256131.1 6 1273
XM_008772746.1 XP_008770968.1 4 1137

View Full Product Details