Gene Transcripts

Gene Symbol
Cog6
Entrez Gene ID
310411
Gene Name
component of oligomeric golgi complex 6
Gene Aliases
Chromosome Location
Chr. 2: 142197566 - 142235054 on Build Rnor_6.0
UniGene ID
Rn.105874
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004262.1 NP_001004262.1 2 274
GenBank mRNA BC079442.1 - 2 274
FQ225755.1 - 65

View Full Product Details