Gene Transcripts
- Gene Symbol
- OR13A1
- Entrez Gene ID
- 79290
- Gene Name
- olfactory receptor family 13 subfamily A member 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 45302294 - 45315608 on Build GRCh38
- UniGene ID
- Hs.532435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131555.1 |
- |
4 |
1960 |
|
View Full Product Details