Gene Transcripts
- Gene Symbol
- PLEKHG7
- Entrez Gene ID
- 440107
- Gene Name
- pleckstrin homology and RhoGEF domain containing G7
- Gene Aliases
- Chromosome Location
- Chr. 12: 92736489 - 92772092 on Build GRCh38
- UniGene ID
- Hs.591013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128530.1 |
- |
9 |
896 |
|
BC137086.1 |
- |
8 |
823 |
9021083 |
View Full Product Details