Gene Transcripts

Gene Symbol
OR4A15
Entrez Gene ID
81328
Gene Name
olfactory receptor family 4 subfamily A member 15
Gene Aliases
OR11-118
Chromosome Location
Chr. 11: 55367884 - 55368918 on Build GRCh38
UniGene ID
Hs.554529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005275.1 NP_001005275.1 1 496

View Full Product Details