Gene Transcripts

Gene Symbol
Wdr34
Entrez Gene ID
296618
Gene Name
WD repeat domain 34
Gene Aliases
Chromosome Location
Chr. 3: 8599251 - 8615329 on Build Rnor_6.0
UniGene ID
Rn.116904
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005542.2 NP_001005542.2 3 549
GenBank mRNA BC082103.1 - 2 333

View Full Product Details