Gene Transcripts

Gene Symbol
OR1K1
Entrez Gene ID
392392
Gene Name
olfactory receptor family 1 subfamily K member 1
Gene Aliases
hg99
Chromosome Location
Chr. 9: 122800123 - 122801073 on Build GRCh38
UniGene ID
Hs.632681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080859.1 NP_543135.1 1 14
GenBank mRNA BC137033.1 - 1 46
BC137034.1 - 1 46 9021030

View Full Product Details