Gene Transcripts

Gene Symbol
Pcdha2
Entrez Gene ID
353234
Gene Name
protocadherin alpha 2
Gene Aliases
Chromosome Location
Chr. 18: 36939205 - 37187661 on Build GRCm38
UniGene ID
Mm.308500
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198117.2 NP_932785.1 1 679
GenBank mRNA AY013760.1 - 1 566
BC030897.1 - 1 679
BC141390.1 - 1 660 9056003

View Full Product Details