Gene Transcripts

Gene Symbol
FCHSD2
Entrez Gene ID
9873
Gene Name
FCH and double SH3 domains 2
Gene Aliases
NWK, SH3MD3
Chromosome Location
Chr. 11: 72836743 - 73142682 on Build GRCh38
UniGene ID
Hs.744959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014824.2 NP_055639.2 5 579
XM_011545409.1 XP_011543711.1 5 461
XM_011545410.2 XP_011543712.1 5 394
XM_017018632.1 XP_016874121.1 5 638
GenBank mRNA AB018312.1 - 4 429
AK297928.1 - 5 463
BC010394.1 - 5 326 4091973
CD358961.1 - 5 593

View Full Product Details