Gene Transcripts

Gene Symbol
Olr56
Entrez Gene ID
293205
Gene Name
olfactory receptor 56
Gene Aliases
Chromosome Location
Chr. 1: 167944448 - 167945398 on Build Rnor_6.0
UniGene ID
Rn.141921
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000130.1 NP_001000130.1 1 135

View Full Product Details