Gene Transcripts
- Gene Symbol
- SPRNP1
- Entrez Gene ID
- 399833
- Gene Name
- shadow of prion protein homolog (zebrafish) pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 133566934 - 133569958 on Build GRCh38
- UniGene ID
- Hs.660426
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126615.1 |
- |
3 |
2254 |
|
View Full Product Details