Gene Transcripts

Gene Symbol
UBOX5
Entrez Gene ID
22888
Gene Name
U-box domain containing 5
Gene Aliases
RNF37, UBCE7IP5, UIP5, hUIP5
Chromosome Location
Chr. 20: 3107573 - 3159910 on Build GRCh38
UniGene ID
Hs.654646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267584.1 NP_001254513.1 3 319
NM_014948.3 NP_055763.1 3 319
NM_199415.2 NP_955447.1 3 319
GenBank mRNA AB020667.1 - 3 287
AK022444.1 - 3 283
AK222570.1 - 3 291
AK222876.1 - 3 244
BC000515.2 - 3 239 2822816
BC046122.1 - 3 279 5554044
CR457089.1 - 2 133
DA101925.1 - 3 319

View Full Product Details