Gene Transcripts

Gene Symbol
Prss27
Entrez Gene ID
287108
Gene Name
protease, serine 27
Gene Aliases
Mpn
Chromosome Location
Chr. 10: 13410899 - 13418260 on Build Rnor_6.0
UniGene ID
Rn.21284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182949.3 NP_891994.3 4 405
GenBank mRNA AB101677.1 - 4 405

View Full Product Details