Gene Transcripts

Gene Symbol
MAST1
Entrez Gene ID
353118
Gene Name
microtubule associated serine/threonine kinase 1
Gene Aliases
Sast
Chromosome Location
Chr. 19: 26026045 - 26053762 on Build Rnor_6.0
UniGene ID
Rn.161721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181089.1 NP_851603.1 17 1940
GenBank mRNA AY227206.1 - 17 1940
AY227207.1 - 17 1940

View Full Product Details