Gene Transcripts

Gene Symbol
SNX13
Entrez Gene ID
23161
Gene Name
sorting nexin 13
Gene Aliases
RGS-PX1
Chromosome Location
Chr. 7: 17790761 - 17940518 on Build GRCh38
UniGene ID
Hs.487648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015132.4 NP_055947.1 4 478
XM_005249672.1 XP_005249729.1 4 488
XM_005249673.4 XP_005249730.1 4 488
XM_011515229.1 XP_011513531.1 4 488
XM_011515230.2 XP_011513532.1 5 535
XM_011515231.2 XP_011513533.1 6 594
XM_011515233.2 XP_011513535.1 4 469
XM_017011893.1 XP_016867382.1 4 488
XM_017011894.1 XP_016867383.1 5 935
XM_017011895.1 XP_016867384.1 4 473
XM_017011896.1 XP_016867385.1 4 471
XM_017011897.1 XP_016867386.1 4 409
GenBank mRNA AB018256.1 - 4 438
AF420470.1 - 4 446
AK001861.1 - 5 442
AK315135.1 - 4 464
AL353943.1 - 4 454
BC045667.1 - 4 448 5298570
BG718588.1 - 4 472
DB050735.1 - 4 489

View Full Product Details