Gene Transcripts

Gene Symbol
CBY1
Entrez Gene ID
25776
Gene Name
chibby family member 1, beta catenin antagonist
Gene Aliases
C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1
Chromosome Location
Chr. 22: 38656653 - 38673850 on Build GRCh38
UniGene ID
Hs.334911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002880.1 NP_001002880.2 6 803
NM_015373.3 NP_056188.1 5 663
GenBank mRNA AB015347.1 - 2 350
AF331041.1 - 5 599
AL050345.1 - 5 642
AL136686.1 - 5 645
BC016139.1 - 5 641 3922707

View Full Product Details