Gene Transcripts
- Gene Symbol
- AHCTF1
- Entrez Gene ID
- 25909
- Gene Name
- AT-hook containing transcription factor 1
- Gene Aliases
- ELYS, MST108, MSTP108, TMBS62
- Chromosome Location
- Chr. 1: 246839098 - 246931978 on Build GRCh38
- UniGene ID
- Hs.300887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001323342.1 |
NP_001310271.1
|
14, 15 |
2204 |
|
NM_001323343.1 |
NP_001310272.1
|
14, 15 |
2204 |
|
NM_015446.4 |
NP_056261.4
|
14, 15 |
1920 |
|
NR_136586.1 |
|
14, 15 |
2204 |
|
XM_006711758.1 |
XP_006711821.1
|
14, 15 |
2033 |
|
XM_011544156.2 |
XP_011542458.2
|
14, 15 |
1899 |
|
XM_011544157.2 |
XP_011542459.2
|
14, 15 |
1899 |
|
GenBank mRNA |
AB059277.1 |
- |
14, 15 |
1920 |
|
AL832213.1 |
- |
4, 5 |
332 |
|
AY157619.1 |
- |
14, 15 |
2038 |
|
- Gene Symbol
- AHCTF1P1
- Entrez Gene ID
- 285116
- Gene Name
- AT-hook containing transcription factor 1 pseudogene 1
- Gene Aliases
- AHCTF1P, ELYS-like
- Chromosome Location
- Chr. 2: 161499067 - 161507902 on Build GRCh38
- UniGene ID
- Hs.470435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_077058.1 |
|
1 |
2020 |
|
View Full Product Details