Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000898.4 | NP_000889.3 | 2 | 305 | |
XM_005272608.3 | XP_005272665.1 | 4 | 405 | ||
XM_017029523.1 | XP_016885012.1 | 4 | 455 | ||
XM_017029524.1 | XP_016885013.1 | 3 | 265 | ||
GenBank mRNA | AK294306.1 | - | 3 | 223 | |
AK298942.1 | - | 2 | 242 | ||
AK310458.1 | - | 2 | 277 | ||
AK312679.1 | - | 2 | 280 | ||
AU127841.1 | - | 2 | 278 | ||
BC022494.1 | - | 2 | 251 | 4800208 | |
DC326844.1 | - | 2 | 306 | ||
M69177.1 | - | 2 | 206 | ||
S62734.1 | - | 2 | 129 |