Gene Transcripts

Gene Symbol
REXO2
Entrez Gene ID
25996
Gene Name
RNA exonuclease 2
Gene Aliases
CGI-114, REX2, RFN, SFN
Chromosome Location
Chr. 11: 114439386 - 114450279 on Build GRCh38
UniGene ID
Hs.677190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015523.3 NP_056338.2 2 305
GenBank mRNA AF151872.1 - 2 256
AK223200.1 - 2 220
AK312042.1 - 2 234
AL110239.1 - 2 239
BC003502.1 - 2 207
BC105024.1 - 2 226 8144027
BC105026.1 - 2 226 8144029
BC107887.1 - 215 6663083
BC143701.1 - 2 229 9052215
CR533500.1 - 2 162
DB471224.1 - 2 306

View Full Product Details