Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000779.3 | NP_000770.2 | 5 | 614 | |
NM_001099772.1 | NP_001093242.1 | 5 | 614 | ||
NM_001319161.1 | NP_001306090.1 | 4 | 569 | ||
NM_001319162.1 | NP_001306091.1 | 3 | 427 | ||
NM_001319163.1 | NP_001306092.1 | 3 | 427 | ||
NR_135003.1 | 5 | 614 | |||
XM_011540832.1 | XP_011539134.1 | 5 | 617 | ||
XM_011540833.1 | XP_011539135.1 | 4 | 249 | ||
XM_017000466.1 | XP_016855955.1 | 4 | 521 | ||
GenBank mRNA | AF491285.1 | - | 5 | 530 | |
AK225576.1 | - | 5 | 566 | ||
AK225578.1 | - | 5 | 566 | ||
AK300921.1 | - | 3 | 427 | ||
AK300941.1 | - | 3 | 276 | ||
AK307715.1 | - | 5 | 688 | ||
AK313170.1 | - | 5 | 566 | ||
AY064484.1 | - | 5 | 530 | ||
AY064485.1 | - | 5 | 530 | ||
AY064486.1 | - | 5 | 530 | ||
AY151048.1 | - | 5 | 530 | ||
AY151049.1 | - | 4 | 485 | ||
BC017758.1 | - | 5 | 566 | 4693225 | |
CD013981.1 | - | 5 | 548 | ||
DC358497.1 | - | 3 | 379 | ||
J02871.1 | - | 5 | 542 | ||
X16699.1 | - | 5 | 530 |