Gene Transcripts
- Gene Symbol
- CEP83
- Entrez Gene ID
- 51134
- Gene Name
- centrosomal protein 83
- Gene Aliases
- CCDC41, NPHP18, NY-REN-58
- Chromosome Location
- Chr. 12: 94268832 - 94459991 on Build GRCh38
- UniGene ID
- Hs.279209
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF155115.1 |
- |
9 |
1332 |
|
View Full Product Details