Gene Transcripts

Gene Symbol
NOL7
Entrez Gene ID
51406
Gene Name
nucleolar protein 7
Gene Aliases
C6orf90, PQBP3, RARG-1, dJ223E5.2
Chromosome Location
Chr. 6: 13615327 - 13632470 on Build GRCh38
UniGene ID
Hs.306242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317724.1 NP_001304653.1 6, 7 654
NM_016167.4 NP_057251.2 6, 7 654
GenBank mRNA AF172066.3 - 6, 7 654
AK075301.1 - 6, 7 637
BC023517.2 - 6, 7 629 4298095
BC062683.1 - 6, 7 650 6499123
BU071935.1 - 6, 7 407

View Full Product Details