Gene Transcripts
- Gene Symbol
- HP1BP3
- Entrez Gene ID
- 50809
- Gene Name
- heterochromatin protein 1 binding protein 3
- Gene Aliases
- HP1-BP74, HP1BP74
- Chromosome Location
- Chr. 1: 20742677 - 20787809 on Build GRCh38
- UniGene ID
- Hs.142442
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016287.3 |
NP_057371.2
|
2, 3 |
247 |
|
XM_005245875.4 |
XP_005245932.1
|
2, 3 |
822 |
|
XM_005245876.3 |
XP_005245933.1
|
2, 3 |
355 |
|
XM_005245877.4 |
XP_005245934.1
|
2, 3 |
683 |
|
XM_005245878.4 |
XP_005245935.1
|
2, 3 |
619 |
|
XM_005245879.4 |
XP_005245936.1
|
1, 2 |
146 |
|
XM_011541532.1 |
XP_011539834.1
|
2, 3 |
339 |
|
XM_011541533.1 |
XP_011539835.1
|
2, 3 |
536 |
|
XM_011541534.2 |
XP_011539836.1
|
1, 2 |
156 |
|
XM_017001393.1 |
XP_016856882.1
|
2, 3 |
302 |
|
XM_017001394.1 |
XP_016856883.1
|
2, 3 |
667 |
|
GenBank mRNA |
AF113534.1 |
- |
1, 2 |
177 |
|
AK023129.1 |
- |
2, 3 |
247 |
|
AK291252.1 |
- |
2, 3 |
225 |
|
AK304065.1 |
- |
1, 2 |
123 |
|
BC032139.2 |
- |
2, 3 |
334 |
|
BE247224.1 |
- |
1, 2 |
219 |
|
View Full Product Details