Gene Transcripts

Gene Symbol
METRNL
Entrez Gene ID
284207
Gene Name
meteorin like, glial cell differentiation regulator
Gene Aliases
Chromosome Location
Chr. 17: 83079691 - 83095126 on Build GRCh38
UniGene ID
Hs.378821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004431.2 NP_001004431.1 2 376
XM_017024523.1 XP_016880012.1 2 725
GenBank mRNA AK093748.1 - 1 947
BC050568.1 - 1 106
BC082252.1 - 2 376 6301543
DR760693.1 - 1 96

View Full Product Details