Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr. 15: 42158701 - 42208331 on Build GRCh38
UniGene ID
Hs.88025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301138.1 NP_001288067.1 12 1219
NM_015289.3 NP_056104.2 11 1186
XM_011521403.2 XP_011519705.1 12 1188
XM_011521404.2 XP_011519706.1 11 1191
XM_017022036.1 XP_016877525.1 12 1187
XM_017022037.1 XP_016877526.1 11 1191
XM_017022038.1 XP_016877527.1 11 1374
XM_017022039.1 XP_016877528.1 11 1159
GenBank mRNA AB018313.2 - 11 1137
AF280814.1 - 12 1046
AF281052.1 - 11 1156
BC015817.2 - 7 746
BC068559.1 - 11 1155 5295166
BX537404.1 - 11 1165

View Full Product Details