Gene Transcripts

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
Chromosome Location
Chr. 17: 18108706 - 18180227 on Build GRCh38
UniGene ID
Hs.462390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016239.3 NP_057323.3 2 1198
XM_011523918.2 XP_011522220.1 1 1558
XM_017024714.1 XP_016880203.1 1 1558
XM_017024715.1 XP_016880204.1 1 1558
GenBank mRNA AB209423.1 - 1 493
AF144094.1 - 2 1198

View Full Product Details