Gene Transcripts

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
Chromosome Location
Chr. 11: 30863603 - 30992686 on Build GRCh38
UniGene ID
Hs.530438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020869.3 NP_065920.2 12 1801
GenBank mRNA AB040926.1 - 2 327
BX647348.1 - 12 1801

View Full Product Details