Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135040.2 | NP_001128512.1 | 9 | 1520 | |
NM_001135041.2 | NP_001128513.1 | 7 | 1243 | ||
NM_001190836.1 | NP_001177765.1 | 9 | 1463 | ||
NM_001190837.1 | NP_001177766.1 | 11 | 1559 | ||
NM_004082.4 | NP_004073.2 | 12 | 1580 | ||
NM_023019.3 | NP_075408.1 | 7 | 1243 | ||
NR_033935.1 | 9 | 1463 | |||
GenBank mRNA | AB209625.1 | - | 4 | 1483 | |
AK297286.1 | - | 9 | 1215 | ||
AK314352.1 | - | 9 | 1464 | ||
AK315014.1 | - | 7 | 1035 | ||
BC071583.1 | - | 7 | 1160 | 4817849 | |
BX640799.1 | - | 12 | 1359 | ||
BX647925.1 | - | 9 | 1224 | ||
BX647972.1 | - | 9 | 1243 | ||
X98801.1 | - | 10 | 1217 |