Gene Transcripts
- Gene Symbol
- DIAPH1
- Entrez Gene ID
- 1729
- Gene Name
- diaphanous related formin 1
- Gene Aliases
- DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
- Chromosome Location
- Chr. 5: 141515021 - 141619055 on Build GRCh38
- UniGene ID
- Hs.529451
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001079812.2 |
NP_001073280.1
|
11, 12 |
1397 |
|
NM_001314007.1 |
NP_001300936.1
|
12, 13 |
1424 |
|
NM_005219.4 |
NP_005210.3
|
12, 13 |
1424 |
|
XM_011537572.2 |
XP_011535874.1
|
11, 12 |
1365 |
|
XM_011537573.2 |
XP_011535875.1
|
12, 13 |
1232 |
|
GenBank mRNA |
AB209482.1 |
- |
12, 13 |
1365 |
|
AF051782.1 |
- |
11, 12 |
1256 |
|
AK023345.1 |
- |
5, 6 |
607 |
|
AK307822.1 |
- |
8, 9 |
856 |
|
AY363395.1 |
- |
12, 13 |
1283 |
|
BC117257.1 |
- |
11, 12 |
1274 |
40125808 |
BC143413.1 |
- |
12, 13 |
1301 |
9051921 |
BC143414.1 |
- |
12, 13 |
1301 |
|
DQ067453.1 |
- |
11, 12 |
1271 |
|
View Full Product Details