Gene Transcripts

Gene Symbol
ZCCHC17
Entrez Gene ID
51538
Gene Name
zinc finger CCHC-type containing 17
Gene Aliases
HSPC251, PS1D, pNO40
Chromosome Location
Chr. 1: 31296982 - 31365764 on Build GRCh38
UniGene ID
Hs.524094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282566.1 NP_001269495.1 2, 3 298
NM_001282567.1 NP_001269496.1 2, 3 160
NM_001282568.1 NP_001269497.1 3, 4 419
NM_001282569.1 NP_001269498.1 4, 5 351
NM_001282570.1 NP_001269499.1 2, 3 160
NM_001282572.1 NP_001269501.1 3, 4 281
NM_001282573.1 NP_001269502.1 4, 5 351
NM_001282574.1 NP_001269503.1 3, 4 281
NM_016505.3 NP_057589.2 3, 4 281
XM_006710681.3 XP_006710744.1 4, 5 425
XM_011541567.2 XP_011539869.1 3, 4 297
GenBank mRNA AF151077.1 - 3, 4 210
AF151085.1 - 3, 4 239
AF247661.2 - 2, 3 150
AK001929.1 - 3, 4 231
AK022506.1 - 3, 4 268
AK024049.1 - 3, 4 257
AK302248.1 - 4, 5 301
AK302343.1 - 2, 3 106
AK307478.1 - 2, 3 137
BC000685.2 - 3, 4 319
BC007446.2 - 3, 4 217 3009823
BC050609.1 - 3, 4 221 6527025
BF309574.1 - 3, 4 232
CN423139.1 - 3, 4 218
DA737244.1 - 4, 5 301
DC366453.1 - 3, 4 419

View Full Product Details