Gene Transcripts
- Gene Symbol
- RALY
- Entrez Gene ID
- 22913
- Gene Name
- RALY heterogeneous nuclear ribonucleoprotein
- Gene Aliases
- HNRPCL2, P542
- Chromosome Location
- Chr. 20: 33993652 - 34084884 on Build GRCh38
- UniGene ID
- Hs.136947
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007367.3 |
NP_031393.2
|
6, 7 |
1180 |
|
NM_016732.2 |
NP_057951.1
|
7, 8 |
1228 |
|
XM_005260334.4 |
XP_005260391.1
|
6, 7 |
1143 |
|
XM_005260336.4 |
XP_005260393.1
|
5, 6 |
1095 |
|
XM_011528694.2 |
XP_011526996.1
|
7, 8 |
786 |
|
XM_011528695.2 |
XP_011526997.1
|
8, 9 |
1338 |
|
XM_017027731.1 |
XP_016883220.1
|
6, 7 |
1143 |
|
GenBank mRNA |
AF148457.1 |
- |
7, 8 |
954 |
|
AK222915.1 |
- |
7, 8 |
914 |
|
AK226085.1 |
- |
6, 7 |
1079 |
|
AK291054.1 |
- |
6, 7 |
751 |
|
AK291079.1 |
- |
7, 8 |
805 |
|
AY635580.1 |
- |
2, 3 |
207 |
|
BC103753.1 |
- |
7, 8 |
975 |
6182358 |
BC105018.1 |
- |
7, 8 |
823 |
8144021 |
L38696.1 |
- |
6, 7 |
784 |
|
View Full Product Details