Gene Transcripts

Gene Symbol
UBQLN3
Entrez Gene ID
50613
Gene Name
ubiquilin 3
Gene Aliases
TUP-1
Chromosome Location
Chr. 11: 5507300 - 5509993 on Build GRCh38
UniGene ID
Hs.189184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017481.2 NP_059509.1 2 193
XM_011520144.2 XP_011518446.1 2 294
XM_011520145.2 XP_011518447.1 2 263
GenBank mRNA AF230481.1 - 2 200
BC036743.1 - 2 208 5167911

View Full Product Details