Gene Transcripts

Gene Symbol
TMEM26
Entrez Gene ID
219623
Gene Name
transmembrane protein 26
Gene Aliases
Chromosome Location
Chr. 10: 61406642 - 61453450 on Build GRCh38
UniGene ID
Hs.623955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178505.7 NP_848600.2 6 4755
NR_134507.1 7 2422
GenBank mRNA AK125611.1 - 7 2210
CR749606.1 - 6 4755

View Full Product Details