Gene Transcripts

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
Chromosome Location
Chr. 10: 126424997 - 126671038 on Build GRCh38
UniGene ID
Hs.587663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004298.2 NP_001004298.2 3 1261
XM_005269491.1 XP_005269548.1 3 1331
XM_011539216.1 XP_011537518.1 4 1989
XM_011539217.2 XP_011537519.1 4 1684
XM_011539220.2 XP_011537522.1 4 1229
XM_011539222.2 XP_011537524.1 4 1351
XM_011539223.2 XP_011537525.1 3 2039
XM_011539224.2 XP_011537526.1 3 1185
XM_011539226.1 XP_011537528.1 3 1184
XM_011539227.2 XP_011537529.1 3 1517
XM_011539230.2 XP_011537532.1 1 924
XM_017015634.1 XP_016871123.1 4 1989
XM_017015635.1 XP_016871124.1 4 1989
GenBank mRNA AF435960.1 - 1 59
AK057500.1 - 3 1261
AK093093.1 - 5 1757
AK297577.1 - 4 1461
AK308357.1 - 4 1348
AK309217.1 - 4 1372
BC140898.1 - 3 1248 9021766
BC146869.1 - 3 1248 9056797

View Full Product Details