Gene Transcripts

Gene Symbol
G0s2
Entrez Gene ID
289388
Gene Name
G0/G1switch 2
Gene Aliases
Chromosome Location
Chr. 13: 112004140 - 112005052 on Build Rnor_6.0
UniGene ID
Rn.1040
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009632.1 NP_001009632.1 2 566
GenBank mRNA BC088248.1 - 2 566

View Full Product Details