Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009672.1 | NP_001009672.1 | 5 | 552 | |
GenBank mRNA | BC088322.1 | - | 5 | 552 | |
FQ214927.1 | - | 5 | 622 | ||
FQ215001.1 | - | 5 | 632 | ||
FQ215643.1 | - | 5 | 597 | ||
FQ224870.1 | - | 5 | 621 | ||
FQ234204.1 | - | 5 | 624 |