Gene Transcripts

Gene Symbol
Hcls1
Entrez Gene ID
288077
Gene Name
hematopoietic cell specific Lyn substrate 1
Gene Aliases
Chromosome Location
Chr. 11: 66736966 - 66759402 on Build Rnor_6.0
UniGene ID
Rn.15578
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011898.1 NP_001011898.1 11 994
GenBank mRNA BC079081.1 - 11 994

View Full Product Details