Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013926.2 | NP_001013948.2 | 4 | 485 | |
XM_003749789.3 | XP_003749837.1 | 4 | 436 | ||
XM_006236804.2 | XP_006236866.1 | 5 | 1142 | ||
XM_006236805.2 | XP_006236867.1 | 5 | 637 | ||
XM_006236808.2 | XP_006236870.1 | 5 | 507 | ||
XM_008763062.1 | XP_008761284.1 | 6 | 1179 | ||
GenBank mRNA | BC087060.1 | - | 5 | 500 | |
CK475733.1 | - | 4 | 485 | ||
CV112449.1 | - | 4 | 413 | ||
FQ234910.1 | - | 4 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006236742.2 | XP_006236804.1 | 1142 | ||
XM_006236743.2 | XP_006236805.1 | 637 | |||
XM_006236745.2 | XP_006236807.1 | 507 | |||
XM_006236746.2 | XP_006236808.1 | 421 | |||
XM_008763108.1 | XP_008761330.1 | 521 | |||
XM_008763109.1 | XP_008761331.1 | 1179 |