Gene Transcripts

Gene Symbol
Il17f
Entrez Gene ID
301291
Gene Name
interleukin 17F
Gene Aliases
Chromosome Location
Chr. 9: 26885702 - 26897219 on Build Rnor_6.0
UniGene ID
Rn.34022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015011.1 NP_001015011.1 5 1174
GenBank mRNA BC091568.1 - 5 1174

View Full Product Details