Gene Transcripts

Gene Symbol
Wbscr17
Entrez Gene ID
288611
Gene Name
Williams-Beuren syndrome chromosome region 17
Gene Aliases
Chromosome Location
Chr. 12: 28382331 - 29268453 on Build Rnor_6.0
UniGene ID
Rn.214637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025112.1 NP_001020283.1 3 437
XM_008769132.1 XP_008767354.1 3 305
GenBank mRNA AB040671.1 - 3 437

View Full Product Details