Gene Transcripts

Gene Symbol
SLC22A20
Entrez Gene ID
440044
Gene Name
solute carrier family 22 member 20
Gene Aliases
Oat6
Chromosome Location
Chr. 11: 65213840 - 65243108 on Build GRCh38
UniGene ID
Hs.532372
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011545043.2 XP_011543345.1 6 1257
GenBank mRNA AK131327.1 - 6 1524

View Full Product Details