Gene Transcripts

Gene Symbol
OR6B2
Entrez Gene ID
389090
Gene Name
olfactory receptor family 6 subfamily B member 2
Gene Aliases
OR2-1, OR6B2P
Chromosome Location
Chr. 2: 240029491 - 240030429 on Build GRCh38
UniGene ID
Hs.531343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005853.1 NP_001005853.1 1 829
GenBank mRNA BC137561.1 - 1 889
BC137562.1 - 1 889 9021562

View Full Product Details