Gene Transcripts

Gene Symbol
Vom1r35
Entrez Gene ID
494268
Gene Name
vomeronasal 1 receptor 35
Gene Aliases
V1rd7
Chromosome Location
Chr. 1: 72128355 - 72129269 on Build Rnor_6.0
UniGene ID
Rn.129430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008919.1 NP_001008919.1 1 20
GenBank mRNA AY510332.1 - 1 20

View Full Product Details