Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009831.2 | NP_001009831.1 | 15 | 2511 | |
XM_006240177.1 | XP_006240239.1 | 13 | 2432 | ||
GenBank mRNA | AY581896.2 | - | 15 | 2511 | |
BC092645.1 | - | 14 | 2434 | 7318365 |