Gene Transcripts

Gene Symbol
Zc3h15
Entrez Gene ID
362154
Gene Name
zinc finger CCCH-type containing 15
Gene Aliases
Chromosome Location
Chr. 3: 71020426 - 71040159 on Build Rnor_6.0
UniGene ID
Rn.129005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010963.1 NP_001010963.1 10 1550
GenBank mRNA BC088438.1 - 10 1550
FQ212631.1 - 5 840

View Full Product Details