Gene Transcripts

Gene Symbol
Vps39
Entrez Gene ID
362199
Gene Name
vacuolar protein sorting 39 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 3: 112087213 - 112125301 on Build Rnor_6.0
UniGene ID
Rn.1508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012186.2 NP_001012186.2 8 824
XM_006234782.2 XP_006234844.1 9 868
GenBank mRNA BC087048.1 - 8 803

View Full Product Details